missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Alcohol, puriss., 99 to 100.5% (GC), Honeywell Riedel-de Haën™
Puriss., meets analytical specification of BP, Ph. Eur., NF, 99-100.5% (GC)
Supplier: Honeywell-Riedel-de Haen 2412225L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
| Benzyl Alcohol | |
| 100-51-6 | |
| -16°C to -13°C | |
| 96°C (204.8°F) | |
| 817°F | |
| C7H8O | |
| MFCD00004599,MFCD03792087 | |
| UN3334 | |
| benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
| OCC1=CC=CC=C1 | |
| 108.14 | |
| 244 | |
| 108.14g/mol | |
| 99 to 100.5% (GC) | |
| Liquid |
| Meets analytical specification of Ph.Eur.BP, NF | |
| 203° to 205°C (lit.) | |
| 1.045g/mL at 25°C | |
| ≤0.005% (as SO4) | |
| Glass Bottle | |
| C6H5CH2OH | |
| 2.5 L | |
| 878307 | |
| WVDDGKGOMKODPV-UHFFFAOYSA-N | |
| phenylmethanol | |
| 3.7 (vs air) | |
| CHEBI:17987 | |
| 13.3mmHg (100°C), 3.75mmHg (77°C) | |
| puriss. |
Chemical Identifiers
| 100-51-6 | |
| 108.14 | |
| WVDDGKGOMKODPV-UHFFFAOYSA-N | |
| 244 | |
| phenylmethanol |
| C7H8O | |
| MFCD00004599,MFCD03792087 | |
| benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
| CHEBI:17987 | |
| OCC1=CC=CC=C1 |
Safety and Handling
P261-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338
H302 + H332-H319
EINECSNumber : 202-859-9
RTECSNumber : DN3150000