missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, NF, EP, bioCERTIFIED, 98-100.5%, Spectrum™ Chemical

Catalog No. 18614891
Change view
Click to view available options
Quantity:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18614891 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18614891 Supplier Spectrum Chemical Mfg Cor Supplier No. B820025LTBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Spectrum™ Chemical bioCERTIFIED products are suitable for biopharmaceutical production.

  • Full testing including endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceability
  • Biopharm industry standard CFR 210/211 cGMP

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Acidity To pass test
Boiling Point 205.3°C
Packaging Drum
Refractive Index 1.538-1.541
Quantity 25 L
Residue after Evaporation 0.0005
Residual Solvents To pass test
Percent Purity 98 to 100.5%
Grade NF, EP, bioCERTIFIED
Physical Form Liquid