missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, NF, EP, bioCERTIFIED, 98-100.5%, Spectrum™ Chemical

Catalog No. 18614867
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614867 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18614867 Supplier Spectrum Chemical Mfg Cor Supplier No. B82004LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Spectrum™ Chemical bioCERTIFIED products are suitable for biopharmaceutical production.

  • Full testing including endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceabilitiy
  • Biopharm industry standard CFR 210/211 cGMP

Chemical Identifiers

CAS 100-51-6

Specifications

Acidity To pass test
Packaging Amber Glass Bottle
Refractive Index 1.538-1.541
Quantity 4 L
Residue after Evaporation 0.05%
Residual Solvents To pass test
Percent Purity 98 to 100.5%
Grade NF/EP/bioCERTIFIED