missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, NF, 98-100.5%, Spectrum™ Chemical

Catalog No. 18600913
Change view
Click to view available options
Quantity:
500 mL
4 L
200 L
Packaging:
Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600913 500 mL Drum
18600914 4 L Drum
18600915 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18600913 Supplier Spectrum Chemical Mfg Cor Supplier No. BE158500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

BE158, 100-51-6, C7H8O

Spectrum™ Chemical Benzyl Alcohol, NF is used as an anti-parasite medication used to treat head lice and as a common preservative in many injectable drugs. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

CAS Max % 100%
Boiling Point 205.3°C
Packaging Drum
Refractive Index 1.538-1.541
Quantity 500 mL
Percent Purity 98 to 100.5%
Grade NF
Physical Form Liquid