missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, Chlorine Free (Benzenemethanol), Reagents

Catalog No. p-200020906
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337466 1 L
01337465 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337466 Supplier Labchem Inc Supplier No. C21521001C
Only null left

Benzyl Alcohol, Chlorine Free (Benzenemethanol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym Phenylcarbinol, Phenylmethanol, Benzenemethanol
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Boiling Range 205°C
Melting Point -15.4°C
Color Colorless
pH Neutral
Boiling Point 205°C
Melting Range -15.4°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.044
Grade Reagent
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol, Chlorine Free (Benzenemethanol)
Show More Show Less