missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, ACS Reagent, Reagents

Catalog No. 01337469
Change view
Click to view available options
Quantity:
4 L
15 gal.
55 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337469 15 gal.
01337468 4 L
01337467 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337469 Supplier Labchem Inc Supplier No. C215208015E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Benzyl Alcohol, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym Phenylcarbinol, Phenylmethanol, Benzenemethanol
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Boiling Range 205°C
Melting Point -15.4°C
Color Colorless
pH Neutral
Boiling Point 205°C
Melting Range -15.4°C
Packaging Steel Drum
Quantity 15 gal.
Specific Gravity 1.044
Grade ACS
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol, ACS Reagent
Show More Show Less