Learn More
Benzyl alcohol, ACS, 99+%
CAS: 100-51-6 | C7H8O | 108.14 g/mol
Supplier: Thermo Scientific Chemicals 041218K7
Description
Benzyl alcohol is widely used as a solvent for the dielectrophoretic reconfiguration of nanowires, inks, paints, lacquers and epoxy resin coatings and as a precursor to a variety of esters used in soaps, perfumes and flavoring. It is employed as a local anesthetic which reduces the pain associated with lidocaine injection. It has a various applications in baby products, bath products, soaps and detergents, eye makeup, blushers, cleansing products, make up products as well as hair, nail and skin care products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Benzyl alcohol | |
| -15°C | |
| 205°C | |
| Aromatic | |
| 1.54 | |
| 4 L | |
| 14,1124 | |
| Misicible with water,benzene,methanol,chloroform,ethanol,acetone and ether. | |
| OCC1=CC=CC=C1 | |
| 108.14 | |
| CHEBI:17987 | |
| ≥99% | |
| Liquid |
| 100-51-6 | |
| 1.045 g/mL | |
| 100°C (212°F) | |
| C7H8O | |
| MFCD00004599,MFCD03792087 | |
| 878307 | |
| benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
| WVDDGKGOMKODPV-UHFFFAOYSA-N | |
| phenylmethanol | |
| 244 | |
| 108.14 | |
| ACS |
Chemical Identifiers
| 100-51-6 | |
| 108.14 | |
| WVDDGKGOMKODPV-UHFFFAOYSA-N | |
| 244 | |
| phenylmethanol |
| C7H8O | |
| MFCD00004599,MFCD03792087 | |
| benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
| CHEBI:17987 | |
| OCC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c
H302+H332-H319
EINECSNumber : 202-859-9
RTECSNumber : DN3150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only