missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Alcohol, ACS, 99%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 205.3°C |
| Ignition Residue | 5e-05 |
| Packaging | Steel Can |
| Quantity | 200 L |
| Percent Purity | 0.99 |
| Grade | ACS |
| Physical Form | Liquid |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.