missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, ACS, 99%, Spectrum™ Chemical

Catalog No. 18609050
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609050 20 L Steel Can
18609053 100 mL Steel Can
18609052 500 mL Steel Can
18609056 4 L Steel Can
18609051 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609050 Supplier Spectrum Chemical Mfg Cor Supplier No. B154620LT
Only null left

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

CAS Max % 1
Boiling Point 205.3°C
Ignition Residue 5e-05
Packaging Steel Can
Quantity 20 L
Percent Purity 0.99
Grade ACS
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.