missing translation for 'onlineSavingsMsg'
Learn More

Benzyl alcohol, 99%

CAS: 100-51-6 | C7H8O | 108.14 g/mol

Supplier:  Thermo Scientific Chemicals L0329230

 View more versions of this product

Catalog No. AAL0329230


Only null left
Explore available promotions

Description

Description

Benzyl alcohol is widely used as a solvent for the dielectrophoretic reconfiguration of nanowires, inks, paints, lacquers and epoxy resin coatings and as a precursor to a variety of esters used in soaps, perfumes and flavoring. It is employed as a local anesthetic which reduces the pain associated with lidocaine injection. It has a various applications in baby products, bath products, soaps and detergents, eye makeup, blushers, cleansing products, make up products as well as hair, nail and skin care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl alcohol is widely used as a solvent for the dielectrophoretic reconfiguration of nanowires, inks, paints, lacquers and epoxy resin coatings and as a precursor to a variety of esters used in soaps, perfumes and flavoring. It is employed as a local anesthetic which reduces the pain associated with lidocaine injection. It has a various applications in baby products, bath products, soaps and detergents, eye makeup, blushers, cleansing products, make up products as well as hair, nail and skin care products.

Solubility
Misicible with water, benzene, methanol, chloroform, ethanol, acetone and ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Benzyl alcohol
-15°C
205°C
Aromatic
1.54
250 g
14,1124
Misicible with water,benzene,methanol,chloroform,ethanol,acetone and ether.
OCC1=CC=CC=C1
108.14
CHEBI:17987
99%
100-51-6
1.045
100°C (212°F)
C7H8O
MFCD00004599,MFCD03792087
878307
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
WVDDGKGOMKODPV-UHFFFAOYSA-N
phenylmethanol
244
108.14

Chemical Identifiers

100-51-6
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
244
phenylmethanol
C7H8O
MFCD00004599,MFCD03792087
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
CHEBI:17987
OCC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c

H302+H332-H319

EINECSNumber : 202-859-9

RTECSNumber : DN3150000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only