missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, 99.0% min, ACS, MilliporeSigma™

Catalog No. MBX04833
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MBX04833 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MBX04833 Supplier Technidata Supplier No. BX04833

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Color Colorless
Density 1.05g/cm3 (20°C)
Explosion Limit 1.3 to 13%(V)
Ignition Point 435°C DIN 51794
Assay Percent Range 99.0% min.
Linear Formula C6H5CH2OH
Quantity 4 L
Vapor Pressure 0.03 hPa (20°C)
Formula Weight 18.14
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol, For ACS analysis
Show More Show Less
Recommended Storage Store at +15°C to +25°C