missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-Bromobutyl Ether 96.0+%, TCI America™

$250.28 - $997.25

Chemical Identifiers

CAS 60789-54-0
Molecular Formula C11H15BrO
Molecular Weight (g/mol) 243.144
InChI Key ZQRVGYAMRJVUAK-UHFFFAOYSA-N
Synonym 4-Benzyloxybutyl Bromide, (4-Bromobutoxymethyl)benzene
PubChem CID 4422181
IUPAC Name 4-bromobutoxymethylbenzene
SMILES C1=CC=C(C=C1)COCCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28025G
View Documents
TCI America
B28025G
5 g
Each for $250.28
Only null left
 
B280225G
View Documents
TCI America
B280225G
25 g
Each for $997.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

60789-54-0
243.144
4-Benzyloxybutyl Bromide, (4-Bromobutoxymethyl)benzene
4-bromobutoxymethylbenzene
C11H15BrO
ZQRVGYAMRJVUAK-UHFFFAOYSA-N
4422181
C1=CC=C(C=C1)COCCCCBr

Specifications

60789-54-0
115°C
5 g
ZQRVGYAMRJVUAK-UHFFFAOYSA-N
4-bromobutoxymethylbenzene
4422181
≥96.0% (GC)
Benzyl 4-Bromobutyl Ether
Yellow
C11H15BrO
4-Benzyloxybutyl Bromide, (4-Bromobutoxymethyl)benzene
C1=CC=C(C=C1)COCCCCBr
243.144
243.14
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'