missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 4-Bromobutyl Ether 96.0+%, TCI America™
Supplier: TCI America B280225G
Specifications
| Benzyl 4-Bromobutyl Ether | |
| Yellow | |
| C11H15BrO | |
| 4-Benzyloxybutyl Bromide, (4-Bromobutoxymethyl)benzene | |
| C1=CC=C(C=C1)COCCCCBr | |
| 243.144 | |
| 243.14 | |
| Liquid |
| 60789-54-0 | |
| 115°C | |
| 25 g | |
| ZQRVGYAMRJVUAK-UHFFFAOYSA-N | |
| 4-bromobutoxymethylbenzene | |
| 4422181 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 60789-54-0 | |
| 243.144 | |
| 4-Benzyloxybutyl Bromide, (4-Bromobutoxymethyl)benzene | |
| 4-bromobutoxymethylbenzene |
| C11H15BrO | |
| ZQRVGYAMRJVUAK-UHFFFAOYSA-N | |
| 4422181 | |
| C1=CC=C(C=C1)COCCCCBr |
Safety and Handling
TSCA : No