missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 3-hydroxyphenylacetate, 98%

Catalog No. p-7027706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2467906 5 g
AAB2467914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2467906 Supplier Thermo Scientific Chemicals Supplier No. B2467906

CAS: 295320-25-1 | C15H14O3 | 242.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 295320-25-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD04039768
InChI Key ALFOBMRIXXPJLQ-UHFFFAOYSA-N
Synonym benzyl 3-hydroxyphenylacetate, benzyl 2-3-hydroxyphenyl acetate, acmc-20alpo, benzyl-3-hydroxyphenylacetate, 3-hydroxyphenylacetic acid benzyl ester, phenylmethyl 2-3-hydroxyphenyl acetate, 3-hydroxy-phenyl-acetic acid benzyl ester
PubChem CID 7016416
IUPAC Name benzyl 2-(3-hydroxyphenyl)acetate
SMILES OC1=CC=CC(CC(=O)OCC2=CC=CC=C2)=C1

Specifications

Melting Point 79°C to 81°C
Quantity 5 g
Formula Weight 242.27
Percent Purity 98%
Chemical Name or Material Benzyl 3-hydroxyphenylacetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only