CAS RN 295320-25-1
IUPAC Name:
benzyl 2-(3-hydroxyphenyl)acetate
Synonyms:
benzyl 2-(3-hydroxyphenyl)acetate
Molecular Weight (g/mol):
242.27
Molecular Formula:
C15H14O3
InChi Key:
ALFOBMRIXXPJLQ-UHFFFAOYSA-N
SMILES:
OC1=CC=CC(CC(=O)OCC2=CC=CC=C2)=C1
1
–
1
of
1
results
1
Promotions Available
Benzyl 3-hydroxyphenylacetate, 98%
Benzyl 3-hydroxyphenylacetate, 98%
CAS: 295320-25-1 Molecular Formula: C15H14O3 Molecular Weight (g/mol): 242.27 MDL Number: MFCD04039768 InChI Key: ALFOBMRIXXPJLQ-UHFFFAOYSA-N Synonym: benzyl 3-hydroxyphenylacetate,benzyl 2-3-hydroxyphenyl acetate,acmc-20alpo,benzyl-3-hydroxyphenylacetate,3-hydroxyphenylacetic acid benzyl ester,phenylmethyl 2-3-hydroxyphenyl acetate,3-hydroxy-phenyl-acetic acid benzyl ester PubChem CID: 7016416 IUPAC Name: benzyl 2-(3-hydroxyphenyl)acetate SMILES: OC1=CC=CC(CC(=O)OCC2=CC=CC=C2)=C1