missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoylthiocholine Iodide 98.0+%, TCI America™
Supplier: TCI America B03075G
Specifications
| Benzoylthiocholine Iodide | |
| White | |
| MFCD00059970 | |
| PCGVSJGEHWZOBK-UHFFFAOYSA-M | |
| [2-(benzoylsulfanyl)ethyl]trimethylazanium iodide | |
| 13960252 | |
| ≥98.0% (T) |
| 10561-14-5 | |
| C12H18INOS | |
| 5 g | |
| [I-].C[N+](C)(C)CCSC(=O)C1=CC=CC=C1 | |
| 351.25 | |
| 351.25 | |
| Crystalline Powder |
Chemical Identifiers
| 10561-14-5 | |
| 351.25 | |
| PCGVSJGEHWZOBK-UHFFFAOYSA-M | |
| [2-(benzoylsulfanyl)ethyl]trimethylazanium iodide |
| C12H18INOS | |
| MFCD00059970 | |
| 13960252 | |
| [I-].C[N+](C)(C)CCSC(=O)C1=CC=CC=C1 |
Safety and Handling
TSCA : No