missing translation for 'onlineSavingsMsg'
Learn More

Benzoylthiocholine Iodide 98.0+%, TCI America™

Catalog No. p-7118406
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B03071G 1 g
B03075G 5 g
B030725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B03071G Supplier TCI America Supplier No. B03071G
Only null left

Chemical Identifiers

CAS 10561-14-5
Molecular Formula C12H18INOS
Molecular Weight (g/mol) 351.25
MDL Number MFCD00059970
InChI Key PCGVSJGEHWZOBK-UHFFFAOYSA-M
PubChem CID 13960252
IUPAC Name [2-(benzoylsulfanyl)ethyl]trimethylazanium iodide
SMILES [I-].C[N+](C)(C)CCSC(=O)C1=CC=CC=C1

Specifications

Color White
Quantity 1 g
Formula Weight 351.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoylthiocholine Iodide