missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl Peroxide (wetted with ca. 25% Water) 75.0+%, TCI America™

Catalog No. B315225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B315225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B315225G Supplier TCI America Supplier No. B315225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94-36-0
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00003071
InChI Key OMPJBNCRMGITSC-UHFFFAOYSA-N
Synonym benzoyl peroxide, dibenzoyl peroxide, peroxide, dibenzoyl, benzoperoxide, benzoyl superoxide, acetoxyl, lucidol, benoxyl, panoxyl, benzol peroxide
PubChem CID 7187
ChEBI CHEBI:82405
IUPAC Name benzoyl benzenecarboperoxoate
SMILES C1=CC=C(C=C1)C(=O)OOC(=O)C2=CC=CC=C2

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 25 g
UN Number 3104
Formula Weight 242.23
Percent Purity ≥75.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoyl Peroxide (wetted with ca. 25% Water)
EINECSNumber (3)-1349
RTECSNumber DM8575000
TSCA Yes
Recommended Storage Refrigerator