missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl Isothiocyanate 97.0+%, TCI America™

Catalog No. p-7131446
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05015G 5 g
I050125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05015G Supplier TCI America Supplier No. I05015G
Only null left

Chemical Identifiers

CAS 532-55-8
Molecular Formula C8H5NOS
Molecular Weight (g/mol) 163.194
MDL Number MFCD00004815
InChI Key CPEKAXYCDKETEN-UHFFFAOYSA-N
Synonym benzoylthiocarbimide, benzoylisothiocyanate, benzoylisothiocyante, isothiocyanic acid benzoyl ester, unii-u8j7q32b86, n-benzoyl isothiocyanate, benzoic acid, anhydride with hncs, benzoic acid, anhydride with isothiocyanic acid, oxophenylmethanisothiocyanate, 1-benzoyl acetone
PubChem CID 68284
IUPAC Name benzoyl isothiocyanate
SMILES C1=CC=C(C=C1)C(=O)N=C=S

Specifications

Boiling Point 143°C
Quantity 5 g
UN Number 2810
Formula Weight 163.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzoyl Isothiocyanate
EINECSNumber (3)-3243
TSCA No