missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl isothiocyanate, 98%

Catalog No. AC314630050
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314630050 5 g Glass bottle
AC314630500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC314630050 Supplier Thermo Scientific Chemicals Supplier No. 314630050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 532-55-8 | C8H5NOS | 163.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 532-55-8
Molecular Formula C8H5NOS
Molecular Weight (g/mol) 163.2
MDL Number MFCD00004815
InChI Key CPEKAXYCDKETEN-UHFFFAOYSA-N
Synonym benzoylthiocarbimide, benzoylisothiocyanate, benzoylisothiocyante, isothiocyanic acid benzoyl ester, unii-u8j7q32b86, n-benzoyl isothiocyanate, benzoic acid, anhydride with hncs, benzoic acid, anhydride with isothiocyanic acid, oxophenylmethanisothiocyanate, 1-benzoyl acetone
PubChem CID 68284
IUPAC Name benzoyl isothiocyanate
SMILES C1=CC=C(C=C1)C(=O)N=C=S

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.2100g/mL
Boiling Point 128.0°C to 131.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6350 to 1.6370
Linear Formula C6H5CONCS
Quantity 5 g
Beilstein 09, IV, 777
Specific Gravity 1.21
Merck Index 15, 1117
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in chloroform,carbon tetrachloride,,dioxane
Formula Weight 163.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Benzoyl isothiocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Toxic if swallowed.
May cause an allergic skin reaction.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 208-540-

RUO – Research Use Only