missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl isothiocyanate, 98%

Catalog No. AAL0696406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0696406 5 g
AAL0696414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0696406 Supplier Thermo Scientific Chemicals Supplier No. L0696406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 532-55-8 | C8H5NOS | 163.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 532-55-8
Molecular Formula C8H5NOS
Molecular Weight (g/mol) 163.194
MDL Number MFCD00004815
InChI Key CPEKAXYCDKETEN-UHFFFAOYSA-N
Synonym benzoylthiocarbimide, benzoylisothiocyanate, benzoylisothiocyante, isothiocyanic acid benzoyl ester, unii-u8j7q32b86, n-benzoyl isothiocyanate, benzoic acid, anhydride with hncs, benzoic acid, anhydride with isothiocyanic acid, oxophenylmethanisothiocyanate, 1-benzoyl acetone
PubChem CID 68284
IUPAC Name benzoyl isothiocyanate
SMILES C1=CC=C(C=C1)C(=O)N=C=S

Specifications

Density 1.214
Boiling Point 128°C to 131°C (15 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6375
Quantity 5 g
UN Number UN2922
Beilstein 606716
Sensitivity Moisture sensitive
Merck Index 14,1114
Formula Weight 163.2
Percent Purity 98%
Chemical Name or Material Benzoyl isothiocyanate
Show More Show Less
Hazard Category H301-H317-H318
Hazard Statement GHS H Statement
H331-H334-H314-H318-H302-H312
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 208-540-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only