Learn More
Benzoyl chloride, 99.5+%, pure
Benzoyl Chloride, 99.5%, C7H5ClO, CAS Number-98-88-4, unii-vty8706w36, a-chlorobenzaldehyde, benzaldehyde, alpha-chloro, benzoic acid chloride, benzenecarbonyl chloride, benzoic acid, chloride, alpha-chlorobenzaldehyde, benzoylchloride, ccris 802, benzoylchlorid, 2.5L, 98.5% min. (GC), 09, 182 | CAS: 98-88-4 | C7H5ClO | 140.57 g/mol
Supplier: Thermo Scientific Chemicals 105750010
Description
Benzoyl Chloride, 99%, C7H5ClO, CAS Number-98-88-4, unii-vty8706w36, a-chlorobenzaldehyde, benzaldehyde, alpha-chloro, benzoic acid chloride, benzenecarbonyl chloride, benzoic acid, chloride, alpha-chlorobenzaldehyde, benzoylchloride, ccris 802, benzoylchlorid, 2.5L, 98.5% min. (GC), 09, 182
Specifications
| Benzoyl Chloride | |
| 98.5 | |
| -1.0°C | |
| 198.0°C | |
| Authentic | |
| Glass Bottle | |
| 1.5520 to 1.5540 | |
| MFCD00000653 | |
| 1736 | |
| 01,50; 05,24; 13,219 | |
| 15, 1115 | |
| Solubility in water: reacts, 2 mg/mL. Other solubilities: decomposes in alcohols and in water, soluble in ether, benzene; miscible with oils,, carbon disulfide, ccl4 | |
| C1=CC=C(C=C1)C(=O)Cl | |
| 140.57 | |
| 0.0012 Pas (30°C) | |
| 140.57 | |
| Pure |
| 98-88-4 | |
| 100.0 | |
| 1.2100g/mL | |
| 93°C | |
| 98.5% min. (GC) | |
| C7H5ClO | |
| C6H5COCl | |
| 1 L | |
| 09, 182 | |
| 1.21 | |
| benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36 | |
| PASDCCFISLVPSO-UHFFFAOYSA-N | |
| benzoyl chloride | |
| 7412 | |
| CHEBI:82275 | |
| 99.5% | |
| Liquid |
Chemical Identifiers
| 98-88-4 | |
| 140.57 | |
| PASDCCFISLVPSO-UHFFFAOYSA-N | |
| 7412 | |
| benzoyl chloride |
| C7H5ClO | |
| MFCD00000653 | |
| benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36 | |
| CHEBI:82275 | |
| C1=CC=C(C=C1)C(=O)Cl |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Toxic if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 202-710-8
RUO – Research Use Only