missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl chloride, 99.5+%, pure

Catalog No. p-3735581
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105750010 1 L Glass Bottle
AC105750025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105750010 Supplier Thermo Scientific Chemicals Supplier No. 105750010
Only null left

Benzoyl Chloride, 99.5%, C7H5ClO, CAS Number-98-88-4, unii-vty8706w36, a-chlorobenzaldehyde, benzaldehyde, alpha-chloro, benzoic acid chloride, benzenecarbonyl chloride, benzoic acid, chloride, alpha-chlorobenzaldehyde, benzoylchloride, ccris 802, benzoylchlorid, 2.5L, 98.5% min. (GC), 09, 182 | CAS: 98-88-4 | C7H5ClO | 140.57 g/mol

Benzoyl Chloride, 99%, C7H5ClO, CAS Number-98-88-4, unii-vty8706w36, a-chlorobenzaldehyde, benzaldehyde, alpha-chloro, benzoic acid chloride, benzenecarbonyl chloride, benzoic acid, chloride, alpha-chlorobenzaldehyde, benzoylchloride, ccris 802, benzoylchlorid, 2.5L, 98.5% min. (GC), 09, 182

Chemical Identifiers

CAS 98-88-4
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
MDL Number MFCD00000653
InChI Key PASDCCFISLVPSO-UHFFFAOYSA-N
Synonym benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
PubChem CID 7412
ChEBI CHEBI:82275
IUPAC Name benzoyl chloride
SMILES C1=CC=C(C=C1)C(=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -1.0°C
Density 1.2100g/mL
Boiling Point 198.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.5520 to 1.5540
Linear Formula C6H5COCl
Quantity 1 L
UN Number 1736
Beilstein 09, 182
Fieser 01,50; 05,24; 13,219
Specific Gravity 1.21
Merck Index 15, 1115
Solubility Information Solubility in water: reacts, 2 mg/mL. Other solubilities: decomposes in alcohols and in water, soluble in ether, benzene; miscible with oils,, carbon disulfide, ccl4
Viscosity 0.0012 Pas (30°C)
Formula Weight 140.57
Percent Purity 99.5%
Grade Pure
Physical Form Liquid
Chemical Name or Material Benzoyl Chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 202-710-8

RUO – Research Use Only