accessibility menu, dialog, popup

UserName

CAS RN 98-88-4

IUPAC Name: benzoyl chloride
Synonyms: benzoyl chloride
Molecular Weight (g/mol): 140.57
Molecular Formula: C7H5ClO
InChi Key: PASDCCFISLVPSO-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
  • (6)
Molecular Weight (g/mol) 
  • (4)
  • (7)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (9)
  • (2)
  • (3)
  • (6)
  • (4)
  • (7)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (10)
  • (3)
  • (3)