missing translation for 'onlineSavingsMsg'
Learn More

Benzotrifluoride, 99%

CAS: 98-08-8 | C7H5F3 | 146.112 g/mol

$75.03 - $199.48

Chemical Identifiers

CAS 98-08-8
Molecular Formula C7H5F3
Molecular Weight (g/mol) 146.112
MDL Number MFCD00000372
InChI Key GETTZEONDQJALK-UHFFFAOYSA-N
Synonym benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene
PubChem CID 7368
ChEBI CHEBI:36810
IUPAC Name trifluoromethylbenzene
SMILES C1=CC=C(C=C1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2134030
View Documents
Thermo Scientific Chemicals
B2134030
250 g
Each for $75.03
Only null left
 
AAB213400B
View Documents
Thermo Scientific Chemicals
B213400B
1000 g
Each for $199.48
Only null left
 
Description

Description

It is applied as an alternative solvent to dichloromethane, where it has a higher bp. It is used as an intermediate for dyes and pharmaceuticals, as a solvent and a dielectric fluid, a vulcanizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an alternative solvent to dichloromethane, where it has a higher bp. It is used as an intermediate for dyes and pharmaceuticals, as a solvent and a dielectric fluid, a vulcanizing agent.

Solubility
Soluble in water at 20°C0.45g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

98-08-8
146.112
GETTZEONDQJALK-UHFFFAOYSA-N
7368
trifluoromethylbenzene
C7H5F3
MFCD00000372
benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene
CHEBI:36810
C1=CC=C(C=C1)C(F)(F)F

Specifications

98-08-8
1.188
12°C (54°F)
C7H5F3
MFCD00000372
UN2338
14,1110
Soluble in water at 20°C0.45g/L.
C1=CC=C(C=C1)C(F)(F)F
146.112
CHEBI:36810
99%
-29°C
101°C to 103°C
Aromatic
1.4145
250 g
1906908
benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene
GETTZEONDQJALK-UHFFFAOYSA-N
trifluoromethylbenzene
7368
146.11
Benzotrifluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BENZOTRIFLUORIDE

EINECSNumber : 202-635-0

RTECSNumber : XT9450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only