missing translation for 'onlineSavingsMsg'
Learn More

Benzotrifluoride, 99%

CAS: 98-08-8 | C7H5F3 | 146.112 g/mol

Supplier:  Thermo Scientific Chemicals B2134030

 View more versions of this product

Catalog No. AAB2134030


Only null left
Explore available promotions

Description

Description

It is applied as an alternative solvent to dichloromethane, where it has a higher bp. It is used as an intermediate for dyes and pharmaceuticals, as a solvent and a dielectric fluid, a vulcanizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an alternative solvent to dichloromethane, where it has a higher bp. It is used as an intermediate for dyes and pharmaceuticals, as a solvent and a dielectric fluid, a vulcanizing agent.

Solubility
Soluble in water at 20°C0.45g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Benzotrifluoride
-29°C
101°C to 103°C
Aromatic
1.4145
250 g
1906908
benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene
GETTZEONDQJALK-UHFFFAOYSA-N
trifluoromethylbenzene
7368
146.11
98-08-8
1.188
12°C (54°F)
C7H5F3
MFCD00000372
UN2338
14,1110
Soluble in water at 20°C0.45g/L.
C1=CC=C(C=C1)C(F)(F)F
146.112
CHEBI:36810
99%

Chemical Identifiers

98-08-8
146.112
GETTZEONDQJALK-UHFFFAOYSA-N
7368
trifluoromethylbenzene
C7H5F3
MFCD00000372
benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene
CHEBI:36810
C1=CC=C(C=C1)C(F)(F)F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BENZOTRIFLUORIDE

EINECSNumber : 202-635-0

RTECSNumber : XT9450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only