missing translation for 'onlineSavingsMsg'
Learn More

(+)-Benzotetramisole 97.0+%, TCI America™

Catalog No. p-7122858
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3296200MG 200 mg
B32961G 1 g
B32965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B3296200MG Supplier TCI America Supplier No. B3296200MG
Only null left

Chemical Identifiers

CAS 885051-07-0
Molecular Formula C15H12N2S
Molecular Weight (g/mol) 252.335
InChI Key YGCWPCVAVSIFLO-LBPRGKRZSA-N
Synonym (2R)-2,3-Dihydro-2-phenylimidazo[2,1-b]benzothiazole, (+)-BTM
PubChem CID 11557884
IUPAC Name (2R)-2-phenyl-1,2-dihydroimidazo[2,1-b][1,3]benzothiazole
SMILES C1C(N=C2N1C3=CC=CC=C3S2)C4=CC=CC=C4

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 200 mg
Formula Weight 252.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Benzotetramisole