accessibility menu, dialog, popup

UserName

CAS RN 885051-07-0

IUPAC Name: 4-phenyl-7-thia-2,5-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),5,8,10-tetraene
Synonyms: 4-phenyl-7-thia-2,5-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),5,8,10-tetraene
Molecular Weight (g/mol): 252.34
Molecular Formula: C15H12N2S
InChi Key: YGCWPCVAVSIFLO-UHFFFAOYNA-N
SMILES: C1C(N=C2SC3=CC=CC=C3N12)C1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results