missing translation for 'onlineSavingsMsg'
Learn More

(+)-Benzotetramisole 97.0+%, TCI America™

Catalog No. B32961G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
5 g
Catalog No. Quantity
B32961G 1 g
B3296200MG 200 mg
B32965G 5 g
3 options

Catalog No. B32961G

Supplier: TCI America B32961G

Only null left

Chemical Identifiers

CAS 885051-07-0
Molecular Formula C15H12N2S
Molecular Weight (g/mol) 252.335
InChI Key YGCWPCVAVSIFLO-LBPRGKRZSA-N
Synonym (2R)-2,3-Dihydro-2-phenylimidazo[2,1-b]benzothiazole, (+)-BTM
PubChem CID 11557884
IUPAC Name (2R)-2-phenyl-1,2-dihydroimidazo[2,1-b][1,3]benzothiazole
SMILES C1C(N=C2N1C3=CC=CC=C3S2)C4=CC=CC=C4

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 1 g
Formula Weight 252.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Benzotetramisole