missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone 99.0+%, TCI America™

Catalog No. p-7118342
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
B008325G 25 g
B0083500G 500 g
2 options

Catalog No. B008325G

Supplier: TCI America B008325G

Only null left

Chemical Identifiers

CAS 119-61-9
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.222
MDL Number MFCD00003076
InChI Key RWCCWEUUXYIKHB-UHFFFAOYSA-N
Synonym benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone
PubChem CID 3102
ChEBI CHEBI:41308
IUPAC Name diphenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 305°C
Quantity 25 g
UN Number 3077
Formula Weight 182.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzophenone
EINECSNumber (3)-1258&(4)-0125
RTECSNumber DI9950000
TSCA Yes