missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone p-toluenesulfonylhydrazone, 97%

Catalog No. p-7035759
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5348606 5 g
AAH5348614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5348606 Supplier Thermo Scientific Chemicals Supplier No. H5348606
Only null left

CAS: 4545-20-4 | C20H18N2O2S | 350.436 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4545-20-4
Molecular Formula C20H18N2O2S
Molecular Weight (g/mol) 350.436
MDL Number MFCD00381345
InChI Key AVWHDUALELPLAI-UHFFFAOYSA-N
Synonym benzophenone tosylhydrazone, n'-diphenylmethylene-4-methylbenzenesulfonohydrazide, n'-diphenylmethylidene-4-methylbenzene-1-sulfonohydrazide, n'-diphenylmethylidene-4-methylbenzenesulfonohydrazide, benzophenone tosyl hydrazone, benzophenone p-tosylhydrazone, benzophenone p-toluenesulfonylhydrazone, benzophenone p-toluenesulphonylhydrazone, benzophenone tosylhydrazone 97
PubChem CID 274531
IUPAC Name N-(benzhydrylideneamino)-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN=C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 178°C to 182°C
Quantity 5 g
Formula Weight 350.44
Percent Purity 97%
Chemical Name or Material Benzophenone p-toluenesulfonylhydrazone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only