CAS RN 4545-20-4
IUPAC Name:
N'-(diphenylmethylidene)-4-methylbenzene-1-sulfonohydrazide
Synonyms:
N'-(diphenylmethylidene)-4-methylbenzenesulfonohydrazide
Molecular Weight (g/mol):
350.44
Molecular Formula:
C20H18N2O2S
InChi Key:
AVWHDUALELPLAI-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)NN=C(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Benzophenone p-toluenesulfonylhydrazone, 97%
Benzophenone p-toluenesulfonylhydrazone, 97%
CAS: 4545-20-4 Molecular Formula: C20H18N2O2S Molecular Weight (g/mol): 350.436 MDL Number: MFCD00381345 InChI Key: AVWHDUALELPLAI-UHFFFAOYSA-N Synonym: benzophenone tosylhydrazone,n'-diphenylmethylene-4-methylbenzenesulfonohydrazide,n'-diphenylmethylidene-4-methylbenzene-1-sulfonohydrazide,n'-diphenylmethylidene-4-methylbenzenesulfonohydrazide,benzophenone tosyl hydrazone,benzophenone p-tosylhydrazone,benzophenone p-toluenesulfonylhydrazone,benzophenone p-toluenesulphonylhydrazone,benzophenone tosylhydrazone 97 PubChem CID: 274531 IUPAC Name: N-(benzhydrylideneamino)-4-methylbenzenesulfonamide SMILES: CC1=CC=C(C=C1)S(=O)(=O)NN=C(C2=CC=CC=C2)C3=CC=CC=C3