missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone Imine 95.0+%, TCI America™

Catalog No. B19125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19125G 5 g
B191225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19125G Supplier TCI America Supplier No. B19125G

Chemical Identifiers

CAS 1013-88-3
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.238
MDL Number MFCD00001760
InChI Key SXZIXHOMFPUIRK-UHFFFAOYSA-N
Synonym benzophenone imine, benzophenoneimine, benzhydrylideneamine, 1,1-diphenylmethanimine, unii-ejj21na7vi, benzhydrylimine, benzophenonimine, diphenylmethanimineimine, 1,1-diphenylmethylimine, benzenemethanimine, .alpha.-phenyl
PubChem CID 136809
IUPAC Name diphenylmethanimine
SMILES C1=CC=C(C=C1)C(=N)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 g
Formula Weight 181.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzophenone Imine