missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone imine, 95%, Stab.

CAS: 1013-88-3 | C13H11N | 181.238 g/mol

$126.93 - $331.88

Chemical Identifiers

CAS 1013-88-3
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.238
MDL Number MFCD00001760
InChI Key SXZIXHOMFPUIRK-UHFFFAOYSA-N
Synonym benzophenone imine, benzophenoneimine, benzhydrylideneamine, 1,1-diphenylmethanimine, unii-ejj21na7vi, benzhydrylimine, benzophenonimine, diphenylmethanimineimine, 1,1-diphenylmethylimine, benzenemethanimine, .alpha.-phenyl
PubChem CID 136809
IUPAC Name diphenylmethanimine
SMILES C1=CC=C(C=C1)C(=N)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3011006
View Documents
Thermo Scientific Chemicals
H3011006
5 g
Each for $126.93
Only null left
 
AAH3011014
View Documents
Thermo Scientific Chemicals
H3011014
25 g
Each for $331.88
Only null left
 
Description

Description

Useful in the preparation of nitrile ylide dimers.Benzophenone imine as ammonia surrogate. As intermediate material for synthesis of organic chemicals. Catalytic enantioselective alkylation of the benzophenone imine of glycine t-butyl ester using phosphazene (Schwesinger) bases and chiral quaternary ammonium salts yields either enantiomer of amino acid derivatives in excellent yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful in the preparation of nitrile ylide dimers.Benzophenone imine as ammonia surrogate. As intermediate material for synthesis of organic chemicals. Catalytic enantioselective alkylation of the benzophenone imine of glycine t-butyl ester using phosphazene (Schwesinger) bases and chiral quaternary ammonium salts yields either enantiomer of amino acid derivatives in excellent yield.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

1013-88-3
181.238
SXZIXHOMFPUIRK-UHFFFAOYSA-N
136809
C1=CC=C(C=C1)C(=N)C2=CC=CC=C2
C13H11N
MFCD00001760
benzophenone imine, benzophenoneimine, benzhydrylideneamine, 1,1-diphenylmethanimine, unii-ejj21na7vi, benzhydrylimine, benzophenonimine, diphenylmethanimineimine, 1,1-diphenylmethylimine, benzenemethanimine, .alpha.-phenyl
diphenylmethanimine

Specifications

1013-88-3
151°C to 153°C (10 mmHg)
1.618
5 g
Insoluble in water.
C1=CC=C(C=C1)C(=N)C2=CC=CC=C2
181.238
181.23
Benzophenone imine
1.08
C13H11N
MFCD00001760
benzophenone imine, benzophenoneimine, benzhydrylideneamine, 1,1-diphenylmethanimine, unii-ejj21na7vi, benzhydrylimine, benzophenonimine, diphenylmethanimineimine, 1,1-diphenylmethylimine, benzenemethanimine, .alpha.-phenyl
SXZIXHOMFPUIRK-UHFFFAOYSA-N
diphenylmethanimine
136809
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only