missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzophenone Hydrazone 98.0+%, TCI America™
Supplier: TCI America B1163500G
Specifications
| Benzophenone Hydrazone | |
| 99°C | |
| 230°C | |
| MFCD00007624 | |
| benzophenone hydrazone, diphenylmethylene hydrazine, benzophenonehydrazone, methanone, diphenyl-, hydrazone, diphenylmethanone hydrazone, benzophenone, hydrazone, diphenylmethylidene hydrazine, diphenyl ketone hydrazone, benzophenone hydrozone, nsc 43 | |
| C1=CC=C(C=C1)C(=NN)C2=CC=CC=C2 | |
| 196.253 | |
| 196.25 | |
| Crystalline Powder |
| 5350-57-2 | |
| White-Yellow | |
| C13H12N2 | |
| 500 g | |
| QYCSNMDOZNUZIT-UHFFFAOYSA-N | |
| benzhydrylidenehydrazine | |
| 79304 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 5350-57-2 | |
| 196.253 | |
| QYCSNMDOZNUZIT-UHFFFAOYSA-N | |
| 79304 | |
| C1=CC=C(C=C1)C(=NN)C2=CC=CC=C2 |
| C13H12N2 | |
| MFCD00007624 | |
| benzophenone hydrazone, diphenylmethylene hydrazine, benzophenonehydrazone, methanone, diphenyl-, hydrazone, diphenylmethanone hydrazone, benzophenone, hydrazone, diphenylmethylidene hydrazine, diphenyl ketone hydrazone, benzophenone hydrozone, nsc 43 | |
| benzhydrylidenehydrazine |
Safety and Handling
EINECSNumber : (4)-1544
TSCA : Yes