missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone Hydrazone 98.0+%, TCI America™

Catalog No. B116325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B116325G 25 g
B1163500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B116325G Supplier TCI America Supplier No. B116325G

Chemical Identifiers

CAS 5350-57-2
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.253
MDL Number MFCD00007624
InChI Key QYCSNMDOZNUZIT-UHFFFAOYSA-N
Synonym benzophenone hydrazone, diphenylmethylene hydrazine, benzophenonehydrazone, methanone, diphenyl-, hydrazone, diphenylmethanone hydrazone, benzophenone, hydrazone, diphenylmethylidene hydrazine, diphenyl ketone hydrazone, benzophenone hydrozone, nsc 43
PubChem CID 79304
IUPAC Name benzhydrylidenehydrazine
SMILES C1=CC=C(C=C1)C(=NN)C2=CC=CC=C2

Specifications

Melting Point 99°C
Color White-Yellow
Boiling Point 230°C
Quantity 25 g
Formula Weight 196.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Benzophenone Hydrazone
EINECSNumber (4)-1544
TSCA Yes