missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone Dimethylketal 98.0+%, TCI America™

Catalog No. p-7119410
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19995G 5 g
B199925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19995G Supplier TCI America Supplier No. B19995G
Only null left

Chemical Identifiers

CAS 2235-01-0
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.29
MDL Number MFCD00025900
InChI Key NYRVXYOKUZSUDA-UHFFFAOYSA-N
Synonym Dimethoxydiphenylmethane
PubChem CID 75228
IUPAC Name [dimethoxy(phenyl)methyl]benzene
SMILES COC(OC)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 106°C
Color White-Yellow
Boiling Point 290°C
Quantity 5 g
Formula Weight 228.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzophenone Dimethylketal
EINECSNumber (4)-1560
TSCA No