accessibility menu, dialog, popup

UserName

CAS RN 2235-01-0

H3COOCH3

CAS RN 2235-01-0

IUPAC Name: [dimethoxy(phenyl)methyl]benzene
Synonyms: [dimethoxy(phenyl)methyl]benzene
Molecular Weight (g/mol): 228.29
Molecular Formula: C15H16O2
InChi Key: NYRVXYOKUZSUDA-UHFFFAOYSA-N
SMILES: COC(OC)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results