missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzophenone Dimethylketal 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 106°C |
| Color | White-Yellow |
| Boiling Point | 290°C |
| Quantity | 5 g |
| Formula Weight | 228.29 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Benzophenone Dimethylketal |
Safety and Handling
| EINECSNumber | (4)-1560 |
| TSCA | No |