missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Benzonase Nuclease

Catalog No. 712063
Change view
Click to view available options
Quantity:
25 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
712063 25 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 712063 Supplier MilliporeSigma™ Supplier No. 712063

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Concentration 99%
Product Type Cell Lysis Enzyme
Content And Storage -20°C
Quantity 25 kU