missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Anhydride 97.0+%, TCI America™

Catalog No. p-7118338
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B007825G 25 g
B0078100G 100 g
B0078500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B007825G Supplier TCI America Supplier No. B007825G
Only null left

Chemical Identifiers

CAS 93-97-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00003073
InChI Key CHIHQLCVLOXUJW-UHFFFAOYSA-N
Synonym benzoic anhydride, benzoic acid, anhydride, benzoyl anhydride, benzoylbenzoate, benzoic acid anhydride, benzoic acid, 1,1'-anhydride, phenyl anhydride, benzoesaeureanhydrid, unii-9k7x34fov2, phenylcarbonyl benzoate
PubChem CID 7167
ChEBI CHEBI:38815
IUPAC Name benzoyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=CC=C2

Specifications

Melting Point 42°C
Color White
Boiling Point 158°C
Quantity 25 g
Formula Weight 226.23
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoic Anhydride
EINECSNumber (3)-3516
TSCA Yes
Recommended Storage Refrigerator