accessibility menu, dialog, popup

UserName

OOO

CAS RN 93-97-0

IUPAC Name: benzoyl benzoate
Synonyms: benzoic anhydride anhydride benzoic acid benzoylbenzoate benzoic acid, 1,1'-anhydride phenyl anhydride 1,1'-anhydride benzoic acid benzoesaeureanhydrid benzoyl anhydride
Molecular Weight (g/mol): 226.23
Molecular Formula: C14H10O3
InChi Key: CHIHQLCVLOXUJW-UHFFFAOYSA-N
SMILES: O=C(OC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (7)
  • (3)
  • (3)
  • (4)
  • (4)
  • (6)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)