missing translation for 'onlineSavingsMsg'
Learn More

Benzoic anhydride, 98%

Numéro de catalogue. p-7021790
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1426918 50 g
AAA1426930 250 g
AAA142690B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAA1426918 Fournisseur Thermo Scientific Chemicals Code fournisseur A1426918
Il en reste null

CAS: 93-97-0 | C14H10O3 | 226.231 g/mol

Benzoic anhydride is used as a benzoylating agent in the manufacture of pharmaceuticals, dyes and intermediates. It serves as an arylation agent in the Heck reaction. It is also used to get benzoic acid through dehydration. Further, it is used to prepare furan-2-yl-phenyl-methanone, which is similar to Friedel-Crafts acylation reaction. In addition to this, it is used for the synthesis of carboxylic esters and lactones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoic anhydride is used as a benzoylating agent in the manufacture of pharmaceuticals, dyes and intermediates. It serves as an arylation agent in the Heck reaction. It is also used to get benzoic acid through dehydration. Further, it is used to prepare furan-2-yl-phenyl-methanone, which is similar to Friedel-Crafts acylation reaction. In addition to this, it is used for the synthesis of carboxylic esters and lactones.

Solubility
Soluble in alcohol, ether, chloroform, acetone, ethyl acetate, benzene, toluene, glacial acetic acid, acetic anhydride and ethanol. Slightly soluble in benzene and acetone. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.

Identifiants chimiques

CAS 93-97-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00003073
InChI Key CHIHQLCVLOXUJW-UHFFFAOYSA-N
Synonym benzoic anhydride, benzoic acid, anhydride, benzoyl anhydride, benzoylbenzoate, benzoic acid anhydride, benzoic acid, 1,1'-anhydride, phenyl anhydride, benzoesaeureanhydrid, unii-9k7x34fov2, phenylcarbonyl benzoate
PubChem CID 7167
ChEBI CHEBI:38815
IUPAC Name benzoyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=CC=C2

Spécifications

Melting Point 40°C to 44°C
Density 1.199
Boiling Point 360°C
Flash Point >110°C (230°F)
Assay Percent Range ≥98%
Quantity 50 g
Beilstein 516726
Sensitivity Moisture sensitive
Merck Index 14,1092
Solubility Information Soluble in alcohol,ether,chloroform,acetone,ethyl acetate,benzene,toluene,glacial acetic acid,acetic anhydride and ethanol. Slightly soluble in benzene and acetone. Insoluble in water.
Formula Weight 226.23
Percent Purity 98%
Chemical Name or Material Benzoic Anhydride
Afficher plus de résultats Afficher moins de résultats
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 202-291-1
TSCA Yes
missing translation for 'storageNote' Moisture sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only