missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid, FCC, 99.5-100.5%, Spectrum™ Chemical

B1102, 65-85-0, C7H6O2

Supplier:  Spectrum Chemical Mfg Cor B110212KG

 View more versions of this product

Catalog No. 18600909


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Benzoic Acid, FCC is used as a food preservative inhibiting the growth of mold, yeast and some bacteria. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

65-85-0
0.05%
C7H6O2
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzoic acid
99.5 to 100.5%
Crystalline Powder or Lumps
100%
Poly Bottle
12 kg
OC(=O)C1=CC=CC=C1
122.12
FCC

Chemical Identifiers

65-85-0
122.12
benzoic acid
C7H6O2
WPYMKLBDIGXBTP-UHFFFAOYSA-N
OC(=O)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions