missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid, FCC, 99.5-100.5%, Spectrum™ Chemical

Catalog No. p-7157036
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600907 500 g Poly Bottle
18600908 2.5 kg Poly Bottle
18600909 12 kg Poly Bottle
18600910 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600907 Supplier Spectrum Chemical Mfg Cor Supplier No. B1102500GM
Only null left

B1102, 65-85-0, C7H6O2

Spectrum™ Chemical Benzoic Acid, FCC is used as a food preservative inhibiting the growth of mold, yeast and some bacteria. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
IUPAC Name benzoic acid
SMILES OC(=O)C1=CC=CC=C1

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99.5 to 100.5%
Grade FCC
Physical Form Crystalline Powder or Lumps