missing translation for 'onlineSavingsMsg'
Learn More

Benzo[a]phenanthrene (purified by sublimation) 98.0+%, TCI America™

Catalog No. C0339100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0339100MG 100 mg
C03391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0339100MG Supplier TCI America Supplier No. C0339100MG
Only null left

Chemical Identifiers

CAS 218-01-9
Molecular Formula C18H12
Molecular Weight (g/mol) 228.294
MDL Number MFCD00003698
InChI Key WDECIBYCCFPHNR-UHFFFAOYSA-N
Synonym benzo a phenanthrene, 1,2-benzophenanthrene, 1,2-benzphenanthrene, 1,2,5,6-dibenzonaphthalene, benz a phenanthrene, chrysen, rcra waste number u050, rcra waste no. u050
PubChem CID 9171
ChEBI CHEBI:51687
IUPAC Name chrysene
SMILES C1=CC=C2C(=C1)C=CC3=C2C=CC4=CC=CC=C43

Specifications

Melting Point 255°C
Boiling Point 448°C
Quantity 100 mg
UN Number 3077
Formula Weight 228.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[a]phenanthrene (purified by sublimation)
RTECSNumber GC0700000
TSCA Yes