CAS RN 218-01-9
CAS RN 218-01-9
IUPAC Name:
chrysene
Synonyms:
chrysene benz(a)phenanthrene 1,2-benzphenanthrene ccris 161 rcra waste number U050 1,2,5,6-dibenzonaphthalene benzo(a)phenanthrene 1,2-benzophenanthrene
Molecular Weight (g/mol):
228.29
Molecular Formula:
C18H12
InChi Key:
WDECIBYCCFPHNR-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(C=CC3=C2C=CC2=CC=CC=C32)=C1
1
–
1
of
1
results
1
Benzo[a]phenanthrene (purified by sublimation) 98.0+%, TCI America™
CAS: 218-01-9 Molecular Formula: C18H12 Molecular Weight (g/mol): 228.294 MDL Number: MFCD00003698 InChI Key: WDECIBYCCFPHNR-UHFFFAOYSA-N Synonym: benzo a phenanthrene,1,2-benzophenanthrene,1,2-benzphenanthrene,1,2,5,6-dibenzonaphthalene,benz a phenanthrene,chrysen,rcra waste number u050,rcra waste no. u050 PubChem CID: 9171 ChEBI: CHEBI:51687 IUPAC Name: chrysene SMILES: C1=CC=C2C(=C1)C=CC3=C2C=CC4=CC=CC=C43