missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzhydrylphosphonic Acid 98.0+%, TCI America™
$138.61 - $475.52
Chemical Identifiers
| CAS | 92025-81-5 |
|---|---|
| Molecular Formula | C13H13O3P |
| Molecular Weight (g/mol) | 248.218 |
| MDL Number | MFCD00013952 |
| InChI Key | JDRCQIRZJMJGMW-UHFFFAOYSA-N |
| Synonym | (Diphenylmethyl)phosphonic Acid |
| PubChem CID | 195023 |
| IUPAC Name | benzhydrylphosphonic acid |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)P(=O)(O)O |
Chemical Identifiers
| 92025-81-5 | |
| 248.218 | |
| JDRCQIRZJMJGMW-UHFFFAOYSA-N | |
| 195023 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)P(=O)(O)O |
| C13H13O3P | |
| MFCD00013952 | |
| (Diphenylmethyl)phosphonic Acid | |
| benzhydrylphosphonic acid |
Specifications
| 92025-81-5 | |
| White | |
| MFCD00013952 | |
| 3261 | |
| JDRCQIRZJMJGMW-UHFFFAOYSA-N | |
| benzhydrylphosphonic acid | |
| 195023 | |
| ≥98.0% (HPLC) | |
| Benzhydrylphosphonic Acid |
| 235°C | |
| C13H13O3P | |
| 200 mg | |
| (Diphenylmethyl)phosphonic Acid | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)P(=O)(O)O | |
| 248.218 | |
| 248.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No