accessibility menu, dialog, popup

UserName

POOHHO

CAS RN 92025-81-5

IUPAC Name: (diphenylmethyl)phosphonic acid
Synonyms: diphenylmethylphosphonic acid
Molecular Weight (g/mol): 248.22
Molecular Formula: C13H13O3P
InChi Key: JDRCQIRZJMJGMW-UHFFFAOYSA-N
SMILES: OP(O)(=O)C(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results