missing translation for 'onlineSavingsMsg'
Learn More

Benzhydrylphosphonic Acid 98.0+%, TCI America™

Catalog No. B46341G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46341G 1 g
B4634200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46341G Supplier TCI America Supplier No. B46341G

Chemical Identifiers

CAS 92025-81-5
Molecular Formula C13H13O3P
Molecular Weight (g/mol) 248.218
MDL Number MFCD00013952
InChI Key JDRCQIRZJMJGMW-UHFFFAOYSA-N
Synonym (Diphenylmethyl)phosphonic Acid
PubChem CID 195023
IUPAC Name benzhydrylphosphonic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)P(=O)(O)O

Specifications

Melting Point 235°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 248.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Benzhydrylphosphonic Acid